Megger MCT105 Cable Locator
Representative image only.
Megger Mfr # 1013-836 – Item # EW-20027-11
Easily locate both live and dead cables and conductors in walls and underground
  • Trace buried conductors and metal water and heating pipes
  • Noncontact voltage detector ensure user safety
  • Measure voltage up to 400 V
  • Visual and audible alarms indicate signal strength
  • Adjust sensitivity automatically or manually
  • Easily view readings on the backlit LCDs
  • 1 year warranty
$215.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • DC Min (Volts)12
  • DC Max (Volts)400
  • AC Min (Volts)12
  • AC Max (Volts)400
  • Frequency Max (KHz)125
  • DisplayBacklit LCDs with bar graph on receiver
  • Safety RatingCAT III 300 V
  • PowerOne 9 V battery each for transmitter and receiver
  • DescriptionCable Locator
  • Warranty1 year
Conformite Europeenne1 Year Warranty
Key Features
  • Trace buried conductors and metal water and heating pipes
  • Noncontact voltage detector ensure user safety
  • Measure voltage up to 400 V
  • Visual and audible alarms indicate signal strength
  • Adjust sensitivity automatically or manually
  • Easily view readings on the backlit LCDs
  • 1 year warranty
More About this Item
Safely locate, diagnose, test, and measure voltage from electrical cables using the MC105 cable locator. The locator, consisting of a transmitter and a receiver, helps you trace cables and metallic conductors in walls or buried underground. The locator can also be used to identify fuses or breakers on circuits, find breaks or short circuits on cables, and trace metal water and heating pipes.
View signal level, the transmission code, and detected voltage (12, 50, 120, 230, and 400 V AC/DC) on the transmitter LCD. It generates a signal on dead or isolated cables and conductor for easy location. The receiver displays the signal strength, the transmission code, and the presence of mains voltage. Select to set signal sensitivity to automatic or manual adjustment for increased accuracy. It can detect buried cables up to 2.5 m (8.2 ft) in depth. An audible signal and bar graph display notify users to when voltage is detected. The receiver has a built-in flashlight—great when working in dimly lit areas. Both the transmitter and receiver feature auto power-off and low battery indicator.