Megger MFT70 Multifunction Tester
Representative image only.
Megger Mfr # 1007-044 – Item # EW-20026-73
A comprehensive testing solution designed for low voltage electrical installations that meets the standards of NFPA70 and the National Electrical Code
  • Covers all the necessary electrical certification tests for industrial, commercial, and domestic fixed wiring installations, including voltage and frequency measurement, insulation tests, continuity resistance, GFCI testing, ground testing, and more
  • With an IEC 61010 CAT IV 300 V rating, this tester can connect anywhere on the system up to the source transformer, without risk of damage from large transient spikes
  • An intuitive design features color-coded controls, a large backlit display, and audible safety warnings
  • Includes memory and Bluetooth® communications for easy data retrieval and traceability
  • Built for tough environments with IP54 protection, ensuring durability against dust and water
$2,335.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • Safety RatingIEC 61010 CAT IV 300 V
  • DescriptionMultifunction Tester
Includes
A standard 3-wire lead set and a mains test lead and a switched probe for fast and easy testing.
Key Features
  • Covers all the necessary electrical certification tests for industrial, commercial, and domestic fixed wiring installations, including voltage and frequency measurement, insulation tests, continuity resistance, GFCI testing, ground testing, and more
  • With an IEC 61010 CAT IV 300 V rating, this tester can connect anywhere on the system up to the source transformer, without risk of damage from large transient spikes
  • An intuitive design features color-coded controls, a large backlit display, and audible safety warnings
  • Includes memory and Bluetooth® communications for easy data retrieval and traceability
  • Built for tough environments with IP54 protection, ensuring durability against dust and water
More About this Item
This versatile instrument offers a wide range of tests required for industrial, commercial, and domestic fixed wiring installations, simplifying the process of electrical certification. Ease of use is a priority with color-coded rotary dials for test selection, a large backlit display that provides clear readings even in challenging conditions, and audible safety warnings when hazardous voltages are present. Additionally, the MFT70 is designed for efficiency, featuring internally rechargeable batteries for reduced ownership costs and built-in memory with Bluetooth® communications for convenient data retrieval and traceability.
Built for rugged reliability, the MFT70 boasts an IP54 protection rating, safeguarding against dust and water, and offers class-leading input protection, making it resilient in tough environments. With no hidden menus or complicated screens, this intuitive tester simplifies the testing process.