Megger MLM50 Laser Distance Meter
Representative image only.
Megger Mfr # 1013-551 – Item # EW-20026-70
Features single or continuous measurement modes with the ability to track maximum and minimum measurements with ease
  • Accuracy of 1/16th inch (up to 33 ft range)
  • Area and volume calculation
  • Meters, feet and inch ranges
  • 20 measurement memory
  • Front or rear measurement reference
  • Built-in spirit level
$131.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
Includes
Protective belt pouch.
Key Features
  • Accuracy of 1/16th inch (up to 33 ft range)
  • Area and volume calculation
  • Meters, feet and inch ranges
  • 20 measurement memory
  • Front or rear measurement reference
  • Built-in spirit level
More About this Item
The Megger MLM50 Laser Distance Meter is a compact, simple to use, laser distance measuring device. It is housed in a shock absorbent IP54 rated protective case and incorporates soft-touch, sure-click buttons. With a measurement range of up to 164 ft, the MLM50 offers the user either single or continuous measurement modes and the ability to track maximum and minimum measurements quickly and easily.
In addition, the unit can automatically calculate area or volume and has the added bonus of recording the last 20 readings into its internal memory for later recall. Memorized values are retained, even if the batteries are removed or replaced.