Megger Replacement Lead Set for MIT300 Series
Representative image only.
Megger Mfr # 1002-001 – Item # EW-20010-36
  • Red and black leads 1.2 m in length
  • 4 mm right angle plugs on instrument ends
  • 4 mm safety probes and crocodile clips on test ends
  • CAT III 1000 V
  • 1 year warranty
$185.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAccessory Leads for MIT300 Series
  • Warranty1 year
1 Year Warranty
Key Features
  • Red and black leads 1.2 m in length
  • 4 mm right angle plugs on instrument ends
  • 4 mm safety probes and crocodile clips on test ends
  • CAT III 1000 V
  • 1 year warranty
More About this Item
Choose from these quality parts and accessories to meet your application needs.