Megger TDR900 Digital Time Domain Reflectometer/Cable Length Meter
Representative image only.
Megger Mfr # TDR900 – Item # EW-20023-18
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDigital Time Domain Reflectometer/Cable Length Meter
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The Megger TDR900 is an advanced instrument capable of measur-ing cable lengths and finding distance to an open or a short using TimeDomain Reflectometry. It offers exceptional features and measure-ment range spans from 15 feet to 10,000 feet with a minimum resolu-tion of 20 inches. The TDR900 can be used for any cable consisting of at least two in-sulated metallic elements, one of which may be the sheath or shield ofthe cable. The meter has automatic internal matching networks to al-low testing of 25O, 50O, 75O, 100O, 125O, or 150Ocables. (These cor-respond to power, telephone, CATV, and LAN cables.)
  • Full autoranging for measuring the length ofpower, telephony, CATV and LAN cables,provides distance to an open or short
  • Extra large, high resolution backlit LCD
  • Automatic output impedance control of 25,50, 75, 100, 125 or 150 ohms
  • User-friendly menu on screen operation
  • Cable length calibration function
  • Internal library of 39 standard cable types
  • Three year warranty
  • REVIEWS