Meiji Techno MX7520 Metallurgical Binocular Microscope; Brightfield/Darkfield/Incident
Representative image only.
Meiji Techno Mfr # MT7520 – Item # EW-48405-31
View metals, plastics, ceramics, paint, glass, crystals, and semiconductor components
  • Plan Epi optics provide clear images, excellent color rendering, and superior flatness of field
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification5x/10x/20x/50x
  • Maximum Magnification50x
  • Head ConfigurationBinocular
  • ObjectivesPlan Epi BF/DF objectives
  • Eyepiece Magnification10x
  • EyepiecesPaired DIN
  • Stage Dimensions191 x 126 mm
  • Illuminator(s)Brightfield/Darkfield/Incident
  • Power (VAC)100 to 240
  • DescriptionMetallurgical Binocular Microscope; Brightfield/Darkfield/Incident
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Plan Epi optics provide clear images, excellent color rendering, and superior flatness of field
  • Limited Lifetime Warranty
More About this Item
Metallurgical microscopes illuminate the specimen through the objective lens — ideal for specimens that don't transmit light, such as metals. Comfortable Siedentopf viewing heads have 30'' inclined eyetubes with one adjustable diopter and one fixed diopter. The interpupillary distance adjustable from 53 to 75 mm. Ergonomic coaxial focus controls have tension adjustment and a safety autostop. Koehler illumination allows for effective contrast control. The illuminator features field iris, aperature iris, and filter slots. Filter sets are included.
Microscopes with brightfield/darkfield/incident illumination give you the flexibility of incident illumination plus adds the capability of darkfield observation mode with Plan Epi BF/DF objectives. This model features an ultra bright 50 W halogen vertical Koehler illuminator. Polarized light mode included.