Meiji Techno S-4100+FX Universal microscope stand and holder
Representative image only.
Meiji Techno Mfr # S-4100+FX – Item # EW-48404-40
Custom design your own microscope
  • Choose the microscope stand that best suits your application
  • Limited Lifetime Warranty
PRODUCT OPTIONS
$1,043.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Stand TypeBoom stand
  • DescriptionUniversal microscope stand and holder
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Choose the microscope stand that best suits your application
  • Limited Lifetime Warranty
More About this Item
Use the components below to build a microscope that best suits your application. We offer a variety of microscope bodies, stands, eyepieces, and ring lights. Components are interchangeable—mix and match to design your perfect microscope! Choose from basic microscope stand, standard microscope stands, or boom microscope stands.
Order microscope and eyepieces separately.
Boom Microscope Stands are ideal when wide horizontal and vertical viewing are important. Stands have their own focusing slide block and illuminator port, and feature a 20-7/8''L horizontal pole and a 15-3/4''L vertical pole. Easily adjust the height on the vertical pole and set it to wherever you need it with the clamp screw. Base measures 10-7/16''L x 10-7/16''W x 1''H.