Meriam 30EBX25-6-06-1 Ruggedized Well-Type Wall-Mount Manometers, Scale F, 15.2 cm WC
Representative image only.
Meriam Mfr # 30EBX25-6-06-1 – Item # EW-68500-43
Accurate and dependable, as well as highly versatile
  • Unit scale extends the full width of the instrument body and is graduated on each side of the indicating column
  • Scale has a white, nonglare background with black graduations and figures
  • Table- or wall-mount style
SPECS & DESCRIPTION
Specifications & Description
  • Min Pressure (in WC)0
  • Max Pressure (in WC)6
  • Liquid FillWater
  • Wetted Materials303 SS
  • DescriptionRuggedized Well-Type Wall-Mount Manometers, Scale F, 15.2 cm WC
Key Features
  • Unit scale extends the full width of the instrument body and is graduated on each side of the indicating column
  • Scale has a white, nonglare background with black graduations and figures
  • Table- or wall-mount style
More About this Item
Well-type manometers provide fast, direct readings of pressure, vacuum, differential, flow and liquid levels. No adding of distances above and below a zero line. All scales are graduated
to correct for the drop in well level and are full width of the instrument.
Ranges: 6, 12, 20, 30, 40, and 60 inches