Meriam B34686 Hand Pump, 145psi, 1/8" NPT(F)
Representative image only.
Meriam Mfr # B34686 – Item # EW-68459-74
  • Features integral bleed and check valves and vernier adjustment
  • Produces up to 145 psig (dead end service)
  • Process connection 1/8" NPT(F)
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure145 psi
  • Min Pressure0 psi
  • Connections1/8\" NPT(F)
  • DescriptionHand Pump, 145psi, 1/8\" NPT(F)
1 Year Warranty
Key Features
  • Features integral bleed and check valves and vernier adjustment
  • Produces up to 145 psig (dead end service)
  • Process connection 1/8" NPT(F)