Meriam DP2000I MeriCal Digital Pressure Calibrator/Manometer; 0 to 2000" H2O Differential
Representative image only.
Meriam Mfr # DP2000I – Item # EW-68459-61
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.1% reading ±1 count
  • Min Pressure (PSID)0
  • Max Pressure (PSID)72
  • Min Pressure (in WC)0
  • Max Pressure (in WC)2000
  • Unit Of Measurement\" WC, cm WC, psi, \" Hg, mm Hg, kg/cm², kPa, mbar
  • Battery Type0 to 30 VDC
  • DescriptionDigital Manometer, +/-0.1% Accuracy
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Display indicates pressure (or differential pressure) and electronic transducer/transmitter output—simultaneously—in any of eight selectable engineering units. Meri-Cal continues to provide the most cost effective and useful method of precise measurements.
The Meri-Cal may be used for test measurement of pressure or differential pressure and as a calibrator for mechanical recorders, gauges, pneumatic controls and electronic transducers and transmitters.