Meriam M101-DN0028 Digital Manometer/Calibrator, 0-28" WC Differential
Representative image only.
Meriam Mfr # M101-DN0028 – Item # EW-68460-35
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.1% full-scale
  • Min Pressure (in WC)0
  • Max Pressure (in WC)28
  • Process Connection1/8\" NPT(F)
  • Unit Of Measurement\" WC
  • Display5-digit LCD, 0.4\" H
  • Battery TypeFour AA (included)
  • Battery Life>100 hours
  • Media CompatibilityClean, dry, noncorrosive gases
  • DescriptionDigital Manometer/Calibrator, 0-28\" WC Differential
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
  • Lightweight, ergonomic, user friendly
  • 5 digit LCD display with 0.4'' high digits
  • Record mode lets user capture up to 240 readings
  • Digital zero and span adjustment
  • 8 selectable user units
  • Ranges from 28 inH2O to 100 PSI
  • 3xAA battery, field replaceable with >100 hours continuous use
  • Auto power off after 20 minutes
  • Media compatibility; non isolated sensors for clean, dry, non-corrosive gases
  • Low battery indicator
  • Process connection 1/8'' female NPT
  • Protective rubber boot
  • Horizontal bar graph for full scale pressure reference
  • Accuracy ± .50 fso standard