Meriam M200-025070300 Smart Manometer, 0/300psig, +/-0.025% FS Accuracy
Representative image only.
Meriam Mfr # M200-025070300 – Item # EW-68460-23
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.025% full-scale
  • Min Pressure (PSI)0
  • Max Pressure (PSI)300
  • Unit Of Measurementpsi
  • Battery TypeFour AA (included)
  • DescriptionSmart Manometer, 0/300psig, +/-0.025% FS Accuracy
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Meriam's M2 Series smart manometers bring high precision and value to handheld, digital manometer users. The M2 features NIST traceable acuracy of ± 0.025% of F.S., independent of temperature effect. Pressure ranges from 28'' H2O F.S. to 2000 PSIG F.S. are availble.