Meriam M200-050200 Smart Manometer, 200in.H2O, 0.05% M1 Series Single Function Calibrator
Representative image only.
Meriam Mfr # M200-050200 – Item # EW-68460-27
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.05% full-scale
  • Min Pressure (in WC)0
  • Max Pressure (in WC)200
  • Unit Of Measurement\" WC
  • Battery TypeFour AA (included)
  • DescriptionSmart Manometer, 200in.H2O, 0.05% M1 Series Single Function Calibrator
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Meriam's M2 Series smart manometers bring high precision and value to handheld, digital manometer users. The M2 features NIST traceable acuracy of ± 0.025% of F.S., independent of temperature effect. Pressure ranges from 28'' H2O F.S. to 2000 PSIG F.S. are availble.