Mettler Toledo 11121058 PWT Touchscreen, Monochrome Display Terminal
Representative image only.
Mettler Toledo Mfr # 11121058 – Item # EW-11140-43
Add an extra balance display to create a truly ergonomic workspace
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPWT Touchscreen, Monochrome Display Terminal
More About this Item
This auxiliary LCD display with backlighting can be placed up to 10 meters from the balance due to the wireless Bluetooth connection. The extra display can be placed elsewhere on your lab bench, outside a fume cupboard or mounted on a stand or wall.