Mettler Toledo 11132630 Auxiliary Display for XP/XS Precision Balances, Locallan Interface
Representative image only.
Mettler Toledo Mfr # 11132630 – Item # EW-11336-88
Add an extra balance display to create a truly ergonomic workspace
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAuxiliary Display for XP/XS Precision Balances, Locallan Interface
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
This auxiliary LCD display with backlighting can be placed up to 10 meters from the balance due to the wireless Bluetooth connection. The extra display can be placed elsewhere on your lab bench, outside a fume cupboard or mounted on a stand or wall.