Mettler Toledo 11140161 U-Electrode Small for Universal Antistastic Kit
Representative image only.
Mettler Toledo Mfr # 11140161 – Item # EW-11336-54
  • Recommended for discharging small samples, 4" x 4.5" internal
  • Ideal for microbalances
  • 3 month warranty
PRODUCT OPTIONS
$3,065.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionU-Electrode Small for Universal Antistastic Kit
  • Warranty3 month
3 Month Warranty
Key Features
  • Recommended for discharging small samples, 4" x 4.5" internal
  • Ideal for microbalances
  • 3 month warranty
More About this Item
Static and electrostatic charging can cause inaccuracies, greatly affecting sensitive balances like analytical, micro, and semi-micro versions.
Many tare vessels, especially plastic containers, are prone to electrostatic charging. Powdery samples then scatter or stick to the container from the static electricity. If the sample is toxic or harmful, this could be dangerous to the operator.
Discharge/neutralize your samples before weighing to ensure maximum security of the sample and user, and obtain more accurate measurements.
Require univeral power supply.