Mettler Toledo 30460839 Evaporation Trap, 10 mL for XPR Micro Analytical Balances
Representative image only.
Mettler Toledo Mfr # 30460839 – Item # EW-11337-72
Optimize performance of your Mettler Toledo touch XPR and XSR Excellence precision balances
  • Keep your balance running smoothly with these parts and accessories
  • Get accurate pipette testing
  • Minimizes errors caused by evaporation
  • Capacity of 10 mL
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeEvaporation Trap
  • DescriptionEvaporation Trap, 10 mL for XPR Micro Analytical Balances
  • Warranty3 Months
3 Month Warranty
Key Features
  • Keep your balance running smoothly with these parts and accessories
  • Get accurate pipette testing
  • Minimizes errors caused by evaporation
  • Capacity of 10 mL
  • 3 month warranty
More About this Item
Choose from a wide range of accessories suitable for your balance including covers, draft shields, ionizers, and more.