Mettler Toledo 30460857 Filter Weighing Kit for XPR/XSR Analytical Balances
Representative image only.
Mettler Toledo Mfr # 30460857 – Item # EW-11337-76
Optimize performance of your Mettler Toledo touch XPR and XSR Excellence precision balances
  • Keep your balance running smoothly with these parts and accessories
  • Enables fast and easy weighing of filter samples up to 110 mm in dia
  • Special weighing cage allows precise positioning of filters
  • 3 month warranty
PRODUCT OPTIONS
$1,295.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeFilter Weighing Kit
  • DescriptionFilter Weighing Kit for XPR/XSR Analytical Balances
  • Warranty3 Months
3 Month Warranty
Key Features
  • Keep your balance running smoothly with these parts and accessories
  • Enables fast and easy weighing of filter samples up to 110 mm in dia
  • Special weighing cage allows precise positioning of filters
  • 3 month warranty
More About this Item
Choose from a wide range of accessories suitable for your balance including covers, draft shields, ionizers, and more.