Mettler Toledo 63052303 Haug Deionizer, without Warning Light
Representative image only.
Mettler Toledo Mfr # 63052303 – Item # EW-11336-96
No more scattering of powdery samples
  • Electrode removes electrostatic discharge reducing contamination and increasing productivity
  • Place electrode next to balance for best results
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHaug Deionizer, without Warning Light
  • Warranty3 month
3 Month Warranty
Key Features
  • Electrode removes electrostatic discharge reducing contamination and increasing productivity
  • Place electrode next to balance for best results
  • 3 month warranty
More About this Item
This deionizer dissipates static charge from the surrounding area, including your sample. Get accurate, reliable results at a quicker pace. Available with or without a warning light.