Mettler Toledo EG13 pH non-aqueous sensor
Representative image only.
Mettler Toledo Mfr # 89632+52300004 – Item # EW-94190-51
Complete packages designed to simplify key applications
  • Simple Apps-based user interface for easy operation
  • Precise burettes and sensors ensure system accuracy
  • Data easily transfered to PC with optional EasyDirect™ software
  • Non-aqueous sensor with cable and BNC connector
  • Used with Easy pH and Easy Pro titrators
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min pH (pH)0
  • Max pH (pH)14
  • mV Resolution0.1
  • DescriptionpH non-aqueous sensor
  • Warranty1 year
1 Year Warranty
Key Features
  • Simple Apps-based user interface for easy operation
  • Precise burettes and sensors ensure system accuracy
  • Data easily transfered to PC with optional EasyDirect™ software
  • Non-aqueous sensor with cable and BNC connector
  • Used with Easy pH and Easy Pro titrators
  • 1 year warranty
More About this Item
Affordable automatic titration packages are designed with everything you need for acid/base, acidity, precipitation, redox, and Karl Fischer volumetric titrations — the ideal replacement for laboratories still performing manual titrations! Although small in size, these quality titrators provide highly accurate and consistent results.
Operation is simple and intuitive thanks to the smartphone-based Apps-driven user interface; use it to control all major functions. Easily perform basic titrations and receive automatic results from predefined formulas. Setup and tutorial with navigation is available in 14 languages. Diagnostic tools assist with the most common issues to minimize downtime.