Mettler Toledo P-52RUE Dot Matrix Printer
Representative image only.
Mettler Toledo Mfr # 30237290 – Item # EW-94190-30
Replacement parts and accessories that are ideal for your Mettler Toledo Halogen Moisture Analyzer.
  • Printing technolog: Dot matrix
  • Printing speed: Up to 10 mm/s
  • Barcode generation: No
  • Interfaces: Ethernet;RS-232;USB Device
  • Operating languages: English
PRODUCT OPTIONS
$1,225.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTitrator Printer
Key Features
  • Printing technolog: Dot matrix
  • Printing speed: Up to 10 mm/s
  • Barcode generation: No
  • Interfaces: Ethernet;RS-232;USB Device
  • Operating languages: English
  • Character set: IBM/DOS, ANSI/WIN, UTF-8