Mettler Toledo R5 Excellence Refractometer, 100-240 VAC
Representative image only.
Mettler Toledo Mfr # R5 – Item # EW-11335-97
Excellence refractometers provide measurements for many challenging samples with an accuracy up to five decimal places
  • Measures in Brix and refractive index
  • Intuitive One Click™ user interface
  • Set up a range of acceptable limits on your samples for immediate QC results
  • Measurement range of 1.32 to 1.58 refractive index
PRODUCT OPTIONS
$14,410.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Width (")15 5/32
  • Density Resolution (g/cm³)0.00001 nD
  • Height (")15 5/32
  • Density Accuracy±0.00002 nD
  • Width (cm)17.37
  • Width (in)15 5/32
  • DescriptionRefractometer Excellence R5
Includes
Refractometer, external power supply, country-specific power cable, terminal WVGA 7-inch AnaChem, combined water standard (9 mL), three plastic pipettes, user manual, declaration of conformity, and test report.
Key Features
  • Measures in Brix and refractive index
  • Intuitive One Click™ user interface
  • Set up a range of acceptable limits on your samples for immediate QC results
  • Measurement range of 1.32 to 1.58 refractive index
More About this Item
These refractometers master measurements of a wide variety of challenging samples with high repeatability and an accuracy of up to five decimal places. The touch-screen terminal with the intuitive One Click™ user interface minimizes training and operator influence. Set up a range of acceptable limits for your samples; results that fall within this range are color-coded, enabling instant identification of accepted and rejected samples.