Mettler Toledo S975 Multiparameter Meter with Stirrer and Probes for pH, Conductivity, and Dissolved Oxygen

Representative image only.
Mettler Toledo Mfr # 30092989 – Item # EW-41000-23
Intuitive and clear interface improves work flow
  • Large color touch screen and plain language menu options make setup and operation easy
  • Predefined and custom methods begin measurements with a single touch
  • Select your preferred display—all the available details or just the basics
  • Integrated radio-controlled clock ensures measurements are correctly stamped with time and date
  • 1 year warranty
Include Buffers Today!
Oakton® pH Buffer Solution Packs
$39.00 - $405.00USD / Pkg of 3

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display type7\" color TFT touch screen
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002
  • Min ISE0 mg/L
  • Max ISE999,999 mg/L
  • ISE Resolution0.001/0.01/0.1/1
  • ISE Accuracy±0.5%
  • Min Conductivity0.01 mS/cm
  • Max Conductivity1000 mS/cm
  • Conductivity Resolution0.001/0.01/0.1/1
  • Conductivity Accuracy±0.5%
  • CellInLab 731-ISM
  • Min Dissolved Oxygen0 mg/L, 0 %
  • Max Dissolved Oxygen99 mg/L, 600 %
  • Dissolved Oxygen Resolution0.001/0.01/0.1
  • Dissolved Oxygen Accuracy±0.5% full-scale or ±0.1 up to 8 mg/L, ±0.2 from 8 to 20 mg/L, ±10% above 20 mg/L, depending on probe
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature Resolution0.1°C
  • Temperature Accuracy±0.1°C
  • Temperature compensationAutomatic or manual; -30 to 130°C
  • OutputUSB, Ethernet, RS-237
  • Data Logging (points)20,000 measurements
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Length (cm)23.5
  • Width (cm)18.8
  • Height (cm)7.5
  • Width (in)7 7/16
  • Length (in)9 1/4
  • Height (in)3
  • DescriptionMultiparameter Meter with Stirrer and Probes for pH, Conductivity, and Dissolved Oxygen
  • Warranty1 year
1 Year Warranty

Includes

The pH/Ion wth conductivity and DO/BOD come with the InLab Expert Pro-ISM pH electrode, InLab 731-ISM conductivity cell, InLab OptiOx dissolved oxygen probe, and uMix® magnetic stirrer.

Key Features

  • Large color touch screen and plain language menu options make setup and operation easy
  • Predefined and custom methods begin measurements with a single touch
  • Select your preferred display—all the available details or just the basics
  • Integrated radio-controlled clock ensures measurements are correctly stamped with time and date
  • 1 year warranty

More About this Item

These dissolved oxygen meters and kits offer a 7" user-friendly color TFT touch screen display programmed with an intuitive menu navigation system, so there is no need to study the manual before you begin taking measurements. With 17 preset and 50 configurable methods available, you can store common setup parameters and switch quickly from one protocol to the next. Choose between the standard view and uFocus™—standard view provides detailed information on parameters and measurements, while uFocus shows only the current reading.

Security features protect data integrity and prevent tampering or mistakes. User management program grants four levels of authority with PIN, reducing unintentional method changes. Intelligent sensor management (ISM®) probes are automatically recognized by the meter which accesses the stored calibration data, so your readings will be accurate even if you switch probes. Built-in radio-controlled clock stamps data with accurate time and date to comply with GLP and other regulatory agencies.

The OptiOx™ optical dissolved oxygen probe uses RDO technology to provide fast, precise, repeatable results. Oxygen is not consumed, so stirring is not a requirement, and since an optical sensor does not need time to polarize, you can begin measuring immediately upon startup.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat