This item is now discontinued. We recommend the following as suggested replacements: EW-35419-65

Mettler Toledo SevenCompact Duo S213 Benchtop pH/Conductivity Meter Water Kit

Representative image only.
Mettler Toledo Mfr # 30339980 – Item # EW-41000-42
Get fast, accurate pH and conductivity measurements with one-touch operation
  • Automatic buffer recognition system offers flexible and fast calibrations
  • Measure both pH and conductivity simultaneously or independently
  • High-resolution color display makes identifying results easier
  • Supports both analog and digital pH and conductivity sensors
  • IP54 water and dust resistance
  • Compliance with GLP regulations
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display type4.3\" color TFT
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002
  • CalibrationConductivity 1; pH Max. 6
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1/1
  • mV Accuracy±0.1
  • Min Conductivity0.001 µS/cm
  • Max Conductivity1000 mS/cm
  • Conductivity Resolution0.001 µS/cm to 1 mS/cm
  • Conductivity Accuracy±0.5% of reading
  • Min TDS0 mg/L
  • Max TDS1000g/L
  • TDS Resolution0.01 mg/L to 1 g/L
  • TDS Accuracy±0.5% of reading
  • Min Salinity0 psu/ppt
  • Max Salinity80 psu/ppt
  • Salinity Resolution0 to 1 psu/ppt
  • Salinity Accuracy±0.5% of reading
  • Min Resistivity0 Ω-cm
  • Max Resistivity100 MΩ-cm
  • Resistivity Resolution0 Ω-cm to 1 MΩ-cm
  • Resistivity Accuracy±0.5% of reading
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)226
  • Temperature Resolution0.1°C
  • Temperature Accuracy±0.1°C
  • OutputUSB, RS-232
  • Data Logging (points)2000 measurements
  • Length (cm)20.4
  • Width (cm)17.4
  • Height (cm)7.4
  • Width (in)6 27/32
  • Length (in)8
  • Height (in)3
  • Probe IncludedYes
  • DescriptionBenchtop pH/Conductivity Meter Water Kit
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP54 Rated

Includes

Electrode holder, protective cover, test certificate, power cord, InLab Puret Pro-ISM pH sensor, InLab 741-ISM conductivity sensor, pH and conductivity guides, two buffer sachets (for pH 4.01, 7.00, 9.21, and 10.00), and two standard sachets (for 1433 µs/cm and 12.88 mS/cm).

Key Features

  • Automatic buffer recognition system offers flexible and fast calibrations
  • Measure both pH and conductivity simultaneously or independently
  • High-resolution color display makes identifying results easier
  • Supports both analog and digital pH and conductivity sensors
  • IP54 water and dust resistance
  • Compliance with GLP regulations
  • 1 year warranty

More About this Item

This multiparameter meter easily toggles between pH and conductivity measurement modes with a single keypress for immediate results. Measure pH/mV and conductivity simultaneously or independently. You can also measure other parameters such as salinity, resistivity, total dissolved solids (TDS), and conductivity ash. Automatic buffer recognition system allows you to choose the order in which to use buffers when calibrating. The meter features eight predefined buffer sets for pH and 13 predefined standards for conductivity.
A user-friendly interface allows you to quickly and easily set up parameters, toggle between screens and function, and change electrodes. The meter menu displays full sentences in 12 languages without cryptic expressions. The Intelligent Sensor Management (ISM®) eliminates mistakes by automatic identification of the sensor upon connection and ensures the most recent calibration is used for future measurements.
Durable construction with IP54 water and dust resistance, rubber caps for connectors, and transparent in-use cover protects the meter from splashes and spills. Integrated USB and RS232 interfaces allow you to connect various accessories such as a barcode reader, keyboard, USB stick, USB printer, or a PC with EasyDirect™ pH software.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat