Mettler Toledo SevenExcellence S400 Benchtop pH/mV Meter Standard Kit with Magnetic Stirrer

Representative image only.
Mettler Toledo Mfr # 30092983 – Item # EW-41000-05
Improve efficiency and productivity
  • Intuitive plain-language interface and touch screen allow rapid familiarization
  • Predefined and custom methods provide the results you need without the hassle of setup
  • PIN-protected user management functions provide security for compliance with regulatory agencies
  • 1 year warranty

PRODUCT OPTIONS

$3,145.00USD / Each

$166.50 - $248.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Buffers Today!
Oakton® pH Buffer Solution Packs
$41.00 - $350.00USD / Pkg of 3

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display type7" color TFT touch screen
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1/1
  • mV Accuracy±0.1
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature Resolution0.1°C
  • Temperature Accuracy±0.1°C
  • Temperature compensationAutomatic or manual
  • OutputUSB, Ethernet, RS-232
  • Data Logging (points)20,000 measurements
  • Power (VAC)115 / 230
  • Power (Hz)50/60
  • Length (cm)20.4
  • Width (cm)17.4
  • Height (cm)7.4
  • Width (in)6 7/8
  • Length (in)8 1/16
  • Height (in)2 7/8
  • DescriptionBenchtop pH/mV Meter Standard Kit with Magnetic Stirrer
  • Warranty1 year
1 Year WarrantyISO9001 Registered Supplier

Includes

two buffer sachets for pH 4.01, 7.00, 9.21, and 10.00, InLab® Expert Pro-ISM PEEK-body general purpose combination electrode, and the uMix magnetic stirrer.

Key Features

  • Intuitive plain-language interface and touch screen allow rapid familiarization
  • Predefined and custom methods provide the results you need without the hassle of setup
  • PIN-protected user management functions provide security for compliance with regulatory agencies
  • 1 year warranty

More About this Item

These pH meters feature an interface designed to improve work flow. The large touch screen makes navigation easy and the menu structure uses plain language, eliminating confusion during set up. One click will start one of 17 predefined or 60 custom methods, enhancing efficiency and throughput. Select the display that works best for you—uFocus™ shows only the most important data while the standard view shows more detail including setup parameters and measurements.

Security features help you comply with regulatory agency requirements. User management settings and PIN-protected functions prevent tampering with custom methods. Integrated Sensor Management (ISM®) pH electrodes are automatically recognized by the meter and store their own calibration data ensuring every measurement is accurate even if you switch electrodes. Time and date stamps from the radio-controlled clock ensure measurements conform to GLP guidelines.

Included electrode arm smoothly lowers and lifts the electrode vertically with one hand, reducing spills and time between measurements. Modular construction allows you to expand your measurement capabilities as your lab needs evolve. Connect with PCs and peripheral devices using the USB, Ethernet, and RS-232 interfaces.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat