Mettler Toledo V10S Compact Volumetric Karl Fischer Titrator with 4 Shortcuts

Representative image only.
Mettler Toledo Mfr # 30267116 – Item # UX-78907-26
Compact yet full of features to provide accurate water content determination
  • Unique cell design ensures accurate and repeatable measurements
  • Easy-to-understand One Click® touchscreen user interface
  • Automatic sensor and burette recognition
  • Burette with RFID chip saves all relevant titrant data
  • Solvent manager provides safe chemical handling
  • Flexible data export via two USB ports
  • Save up to 4 shortcuts per user and 5 analysis methods
  • 1 year warranty

PRODUCT OPTIONS

$10,500.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Unitsmg, µg, ppm, %, g/kg, mg/g, mg/mL, g/mL, µg/L, µg/mL
  • Display TypeVGA 5.7" color TFT touchscreen
  • Display Resolution640 x 480 pixels
  • OutputUSB (2), Ethernet
  • Data LoggingYes
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Length (in)13 1/8
  • Width (in)8 1/4
  • Height (in)12 1/8
  • Length (cm)33.3
  • Width (cm)21
  • Height (cm)30.8
  • DescriptionV10S Compact Volumetric Karl Fischer Titrator with 4 Shortcuts
  • Warranty1 year

Includes

Titration vessel, 5 mL interchangeable burette set, silicone tubing, solvent manager set, 1 L clear glass bottle, dual platinum pin electrode, triaxial SC LEMO cable, 250 g molecular sieve, glass wool, fittings, caps, touchscreen protective cover, external power supply, and power cable.

Key Features

  • Unique cell design ensures accurate and repeatable measurements
  • Easy-to-understand One Click® touchscreen user interface
  • Automatic sensor and burette recognition
  • Burette with RFID chip saves all relevant titrant data
  • Solvent manager provides safe chemical handling
  • Flexible data export via two USB ports
  • Save up to 4 shortcuts per user and 5 analysis methods
  • 1 year warranty

More About this Item

These compact volumetric Karl Fischer titrators offer everything you need for fast and precise water content determination in an easy-to-use system. Analyze samples with water content between 100 ppm to 100% in single measurements or sample series with statistics. Robust design and easy-to-use interface make these simple to use.

Special titration cell design ensures accurate and repeatable measurements by minimizing ambient moisture interference, resulting in a low background drift value. Use the large color touchscreen to operate the simple-to-use One Click user interface to customize home screens to run your workflows with a single tap. Each user can set their own home screen with customized language selection (English, German, French, Spanish, Italian, Chinese, Russian, Polish, Korean, or Portuguese) and shortcut keys.

Automatic sensor and burette recognition reduces extra data entry steps and helps minimize errors. Burettes feature an RFID chip that contains concentration or lifetime of the titrant and is directly transferred to the titrator. Ensure user safety with the solvent manager. It is fully controlled by the titrator, and offers filling, draining, and solvent exchange without additional user interaction. For secure titrator operation, designate user groups to allow full or limited access. Easily export data in CSV, PDF, or XML files using a memory stick or connect to a printer via the USB port.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat