Mettler Toledo WeightLink 5G Test Weight
Representative image only.
Mettler Toledo Mfr # 30293489 – Item # EW-01030-33
Designed for positive tolerances
  • High grade stainless steel offers uniform consistency and resistance to corrosion
  • Unique identification number on base links to certificate
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Mass ClassClass 1
  • Mass Size (g)5
  • Description 5G Test Weight
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • High grade stainless steel offers uniform consistency and resistance to corrosion
  • Unique identification number on base links to certificate
  • Limited Lifetime Warranty
More About this Item
These high grade stainless steel WeightLink test weights, combined with a data matrix code (DMC) scanner (sold separately), are part of an innovative, fool-proof identification and traceability weight verification system for balance testing. The weights range from 1 to 100 g with a unique identification number imbedded on their base linked to a calibration certificate, and a calibration eData card. They come with a lifetime warranty.