Mettler Toledo WeightLink CarePac Test Weight Set 100G and 2000G
Representative image only.
Mettler Toledo Mfr # 30293472 – Item # EW-01030-28
Designed for positive tolerances
  • High grade stainless steel offers uniform consistency and resistance to corrosion
  • Unique identification number on base links to certificate
  • Calibration certificate included
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Mass ClassClass 1
  • Contents of Set100 g, 2000 g
  • DescriptionTest Weight Set 100G and 2000G
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Includes
Calibration certificate, data matrix code card, and plastic storage case.
Key Features
  • High grade stainless steel offers uniform consistency and resistance to corrosion
  • Unique identification number on base links to certificate
  • Calibration certificate included
  • Limited Lifetime Warranty
More About this Item
These high grade stainless steel WeightLink test weights, combined with a data matrix code (DMC) scanner (sold separately), are part of an innovative, fool-proof identification and traceability weight verification system for balance testing. The weights range from 0.1 to 5000 g with a unique identification number imbedded on their base linked to a calibration certificate, and a calibration eData card. They come with a lifetime warranty, guaranteed positive tolerance, and an easy-to-clean plastic storage box.
The test weights are available in a CarePac which contains two weights for testing at 5% and 100% of your balance capacity in accordance with USP recommendations. An optional third weight can be purchased for specific testing requirements.