Mettler Toledo WeightLink DMC Scanner
Representative image only.
Mettler Toledo Mfr # 30268560 – Item # EW-01030-38
Get guaranteed full traceability of your balance tests
  • Fast and secure—parameters of test weights automatically transfer to balance firmware
  • Validates certificate and weight data before allowing balance test to begin
  • Detailed test report provides documentation of performed balance test
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Description DMC Scanner
  • Warranty1 Year
1 Year WarrantyCanadian Standards Association
Key Features
  • Fast and secure—parameters of test weights automatically transfer to balance firmware
  • Validates certificate and weight data before allowing balance test to begin
  • Detailed test report provides documentation of performed balance test
  • 1 year warranty
More About this Item
Pass your next audit with confidence! The WeightLink™ system is an innovative, foolproof identification and traceability weight verification system for fast and error-free balance testing. It consists of a data matrix code (DMC) scanner, test weights with a unique identification number (UIN) imprinted in their base linked to a calibration certificate, and a calibration eData card.
The DMC scanner, which connects directly to a Mettler Toledo analytical and precision balance via a RS-232 port, scans the eData card and test weight codes and automatically transfers the parameters of the test weights to the balance. The balance firmware validates the certificate and weight data before allowing the balance test to begin. Test results can be printed out with an external printer to provide full documentation of performed balance tests.
The high-grade stainless steel test weights, available from 0.1 to 5000 g, are sold separately. They come with a lifetime warranty and guaranteed positive tolerances.
Compatible with Mettle Toledo XP, XS, XPE, and XSE balances.