Microflex Black Dragon Latex Gloves, X-small, 2000/cs
Representative image only.
Microflex Mfr # BD-1000-PF – Item # EW-86231-55
Fully textured for excellent tactile sensitivity and grip
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)5.1
  • MaterialLatex
  • SizeX-Small
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlack
  • CuffBeaded
  • SterileNo
  • DescriptionLatex Gloves, X-small, 2000/cs
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Powder-free gloves feature a combination of comfort and strength. Black color eliminates distractions from stains and coloring on the glove. Beaded cuff for easy donning and removal. Disposable; for single-use only. Ambidextrous.