Microflex CFG-900-L Latex gloves, large, 1000/cs
Representative image only.
Microflex Mfr # CFG-900-L – Item # EW-86232-87
When comfort, quality, and value are crucial to your needs
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLatex
  • Thickness (mil)5.9
  • SizeLarge
  • Length (in)9 1/2
  • Length (cm)24
  • ColorClear
  • GripTextured
  • CuffBeaded
  • SterileNo
  • DescriptionLatex gloves, large, 1000/cs
Conformite Europeenne
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
These powder-free latex gloves provide maximum sensitivity required when using small instruments. Gloves are fully textured to provide secure grip in wet and dry environments. Beaded cuffs offer improved tear resistance while donning and removal. Ambidextrous.