Microflex DGP350-XL Latex Gloves; size, extra-large, 1000/cs
Representative image only.
Microflex Mfr # DGP350-XL – Item # EW-86232-08
Great elasticity and tactile sensitivity make this an ideal multipurpose glove
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLatex
  • Thickness (mil)6.3
  • SizeX-Large
  • Length (in)9 1/2
  • Length (cm)24
  • ColorWhite
  • CuffBeaded
  • SterileNo
  • DescriptionLatex Gloves; size, extra-large, 1000/cs
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Enjoy comfortable wear, durable protection, a secure grip, and tactile sensitivity. Powder-free to reduce skin irritation. Gloves feature a textured surface for excellent grip — wet or dry. Natural latex ensures superior comfort and resilience. Beaded cuffs improve tear resistance and protect hands. Disposable — for single use only. Ambidextrous.
Diamond Grip Plus™ Latex Gloves feature a fully textured surface to ensure exceptional grip and superb tactile sensitivity for instrument manipulation.