Microflex FreeForm SE Nitrile Gloves, extended cuff, small, 500/cs
Representative image only.
Microflex Mfr # FFE-775-S – Item # EW-86445-24
Non-stick properties resist tape and adhesives
  • Certified to the Emergency Medical Glove Requirements of NFPA 1999
  • Non-stick properties resist tape and adhesives
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)6.3
  • MaterialNitrile
  • SizeSmall
  • Length (in)11 1/2
  • Length (cm)29
  • ColorBlue
  • CuffExtended
  • SterileNo
  • DescriptionNitrile Gloves, extended cuff, small, 500/cs
  • Warranty1 month
1 Month WarrantyConformite Europeenne
Key Features
  • Certified to the Emergency Medical Glove Requirements of NFPA 1999
  • Non-stick properties resist tape and adhesives
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
  • 1 month warranty
More About this Item
The gloves feature a palm thickness of 3.5 mil and a finger thickness of 5.1 mil, ideal for light to mid-duty work. Lightly textured fingers ensure a secure grip in wet and dry conditions. Interior polymer coating assists in quick and easy donning.
The 100% nitrile gloves offer superior comfort and elasticity in a synthetic glove. Unique formulation conforms to the contours of your hands as you move — minimizing unnecessary hand stress and enabling a full range-of-motion. Ideal for use in the food processing, life sciences and healthcare industries.
Standards and certifications include: ASTM D6319, EN 420:2003 + A1:2009, FDA21 CFR 177-2600-US Food Contact Approved, NFPA 1999:2018, and US QSR/FDA 510(k) Medical Examination Grade. Ambidextrous.