Microflex MF300-M Latex Gloves, Medium; 1000/CS
Representative image only.
Microflex Mfr # MF300-M – Item # EW-86231-36
Proven performance that consistently exceeds the standards
  • Medical examination grade protects hands of healthcare professionals
  • Double chlorinated for easy donning and a comfortable feel
  • Chemical resistant
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
  • 1 month warranty
PRODUCT OPTIONS
$179.25USD / Case of 1000
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLatex
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • ColorClear
  • CuffBeaded
  • SterileNo
  • Palm Thickness (mil)6.3
  • PowderedNo
  • DescriptionLatex Gloves; size, medium, 1000/cs
  • Warranty1 month
1 Month Warranty
Key Features
  • Medical examination grade protects hands of healthcare professionals
  • Double chlorinated for easy donning and a comfortable feel
  • Chemical resistant
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
  • 1 month warranty
More About this Item
The gloves are designed to be extra thick at the fingers (7.9 mil) for added protection against rips, snags, and tears. Protects your hands in the most demanding environments. Textured fingers allow you to securely grip tools and materials — wet or dry. Natural latex ensures superior comfort and resilience.
The gloves are ideal for use in the automotive, lab research, life sciences, machinery, equipment and healthcare industries.
Standards and certifications include ASTM D3578, EN 420-2003 + A1:2009, FDA21 CFR 177-2600-US Food Contact Approved, and US QSR/FDA 510(k) Medical Examination Grade. Ambidextrous.
Please Note: This product contains natural rubber latex (latex) which may cause allergic reactions in some users. Safe use of this glove by or on latex sensitized individuals has not been established.