Microflex NPG888-XL Chloroprene gloves, X-large, 1000/cs
Representative image only.
Microflex Mfr # NPG888-XL – Item # EW-86232-93
The integrity of a synthetic with the feel of natural latex
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialChloroprene
  • Thickness (mil)6.7
  • SizeX-Large
  • Length (in)9 1/2
  • Length (cm)24
  • ColorGreen
  • GripTextured
  • CuffBeaded
  • SterileNo
  • DescriptionChloroprene gloves, X-large, 1000/cs
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Powder-free chloroprene gloves reduce the risk of skin irritation — ideal for those who prefer non-latex gloves. They provide high-barrier performance and are textured for a more secure grip. For single use only. Beaded, tear-resistant cuff. Ambidextrous.