Microflex SG375-L Latex Gloves; size, large, 500/cs
Representative image only.
Microflex Mfr # SG375-L – Item # EW-86231-67
Extra thickness and tensile strength make this glove ideal for heavy duty applications
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)14.2
  • MaterialLatex
  • SizeLarge
  • Length (in)11 1/2
  • Length (cm)29
  • ColorBlue
  • CuffBeaded
  • SterileNo
  • DescriptionLatex Gloves; size, large, 500/cs
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Enjoy comfortable wear, durable protection, a secure grip, and tactile sensitivity. Powder-free to reduce skin irritation. Gloves feature a textured surface for excellent grip — wet or dry. Natural latex ensures superior comfort and resilience. Beaded cuffs improve tear resistance and protect hands. Disposable — for single use only. Ambidextrous.
SafeGrip® Latex Gloves are ideal for enhanced protection. They have extended cuffs and an average thickness of no less than 12 mil. Superior tensile strength provides extraordinary resiliency.