Microflex SY911-S Latex Gloves; size, small, 500/cs
Representative image only.
Microflex Mfr # SY911-S – Item # EW-86231-45
Extra thick glove provides added durability for tough tasks
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLatex
  • Thickness (mil)11.8
  • SizeSmall
  • Length (in)11 1/2
  • Length (cm)29
  • ColorClear
  • CuffBeaded
  • SterileNo
  • DescriptionLatex Gloves; size, small, 500/cs
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Enjoy comfortable wear, durable protection, a secure grip, and tactile sensitivity. Powder-free to reduce skin irritation. Gloves feature a textured surface for excellent grip — wet or dry. Natural latex ensures superior comfort and resilience. Beaded cuffs improve tear resistance and protect hands. Disposable — for single use only. Ambidextrous.
Synetron® Latex Gloves feature extended cuffs for extra protection and offer excellent dexterity and tactile sensitivity. Polymer-coated interior makes them easy to put on and take off.