Micropump 81101 Economical drive for A-Mount pump heads, 150 to 4000 rpm, 0 to 24 VDC

Representative image only.
Micropump Mfr # 81101 – Item # EW-07003-90
A variety of low-cost drives for numerous applications
  • Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
  • Choose either an explosion-proof or air-powered model for safe pumping in hazardous areas
  • Hundreds of hours of dependable leakproof service

PRODUCT OPTIONS

$457.00USD / Each
Often Bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Min Motor RPM (rpm)300
  • Max Motor RPM (rpm)4000
  • Motor Enclosure TypePM brush, TENV
  • Power (VDC)0 to 24
  • Power (amps)3
  • Duty Cycle ContinuousYes
  • Max Temperature (° F)140
  • Min Temperature (° F)40
  • Max Temperature (° C)60
  • Min Temperature (° C)4
  • Width (in)3
  • Width (cm)7.6
  • Length (in)4
  • Length (cm)10.2
  • Height (in)3
  • Height (cm)7.6
  • Motor hp0.09
  • DescriptionEconomical drive for A-Mount pump heads, 150 to 4000 rpm, 0 to 24 VDC

Key Features

  • Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
  • Choose either an explosion-proof or air-powered model for safe pumping in hazardous areas
  • Hundreds of hours of dependable leakproof service

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat