Micropump GA Series B-Mount Suction Shoe Gear Pump Head, SS Base, Graphite Gear, PTFE Seal; 0.042 mL/rev
Representative image only.
Micropump Mfr # MI0007 / Z-181 / 82114 – Item # EW-74004-29
Smooth pulseless flow provides precise, leak-free delivery of fluids
  • Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency
  • Cavity style designs with no dynamic seals for applications that cannot tolerate leaks
PRODUCT OPTIONS
$894.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • SeriesGA
  • Flow Rate (mL/rev)0.042
  • Differential Pressure (PSI)40
  • Differential Pressure (bar)2.8
  • Max Motor RPM (rpm)5500
  • Seal MaterialPTFE
  • Body MaterialStainless steel
  • Gear MaterialCarbon graphite
  • Inlet Connection (NPT(F))0
  • Outlet Connection (NPT(F))0
  • Internal BypassNo
  • Duty Cycle ContinuousYes
  • ParticulatesNo
  • Max Temperature (° F)350
  • Min Temperature (° F)-50
  • Max Temperature (° C)177
  • Min Temperature (° C)-46
  • Max Viscosity (cP)1500
  • Min Viscosity (cP)0.5
  • DescriptionGear Pump Head, Suction Shoe Style, Stainless Steel, Carbon Graphite Gear, PTFE Seal, 0.042 mL/rev
Key Features
  • Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency
  • Cavity style designs with no dynamic seals for applications that cannot tolerate leaks
More About this Item
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
Call our Application Specialists for assistance.