Micropump Gear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0.91 ml/rev
Representative image only.
Micropump Mfr # 85464 – Item # EW-74445-25
Complete pumps include head, motor, and integrated drive
  • Variable-speed electronic controller delivers smooth, pulseless flow
  • Static seals reduce maintenance
  • Small envelope-ideal for OEM use
PRODUCT OPTIONS
$2,313.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Flow Rate (mL/rev)0.91
  • Min Flow Rate (mL/min)455
  • Max Flow Rate (LPM)4.55
  • Max Flow Rate (GPM)1.2
  • DescriptionGear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0.91 ml/rev
Includes
2-ft (0.6-m) power cord and screwdriver. Suction shoe service kits include: gears, suction shoe, shafts, scales, and springs. Cavity-style service kits include: gears, bushings, and seals.
Key Features
  • Variable-speed electronic controller delivers smooth, pulseless flow
  • Static seals reduce maintenance
  • Small envelope-ideal for OEM use
More About this Item
Electromagnetically driven, precision-geared pumps provide smooth, accurate, pulseless flow. Torque improvements and compact size are achieved with NdFeB magnets. Control speed by using a 0 to 5 VDC input signal. Drives feature thermal and overload protection.
Choose a system with a suction shoe pump head to self compensate for wear, temperature, and pressure, and to maintain high volumetric efficiency. Cavity-style pump heads deliver higher lift capability and moderate pressure capability; an internal bypass is an integral safety feature of the cavity-style pump heads.