Micropump L22096 A-Mount Air-Driven Pump Drive, 500 to 8000 rpm, 5 to 11 scfm, 50 to 80 psi

Representative image only.
Micropump Mfr # L22096 – Item # EW-07144-97
A variety of low-cost drives for numerous applications
  • Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
  • Choose either an explosion-proof or air-powered model for safe pumping in hazardous areas
  • Hundreds of hours of dependable leakproof service

PRODUCT OPTIONS

$674.25USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Min Motor RPM (rpm)500
  • Max Motor RPM (rpm)8000
  • Motor Enclosure TypeTENV
  • Power (VAC)115 / 230
  • Power (Air)Yes
  • Air Consumption (SCFM)11
  • Air Consumption (m³/hr)18
  • Max Air Inlet Pressure (psig)80
  • Max Air Inlet Pressure (bar)5.5
  • Duty Cycle ContinuousYes
  • Max Temperature (° F)140
  • Min Temperature (° F)40
  • Max Temperature (° C)60
  • Min Temperature (° C)4
  • Width (in)3
  • Width (cm)7.6
  • Length (in)3.75
  • Length (cm)9.5
  • Height (in)5.25
  • Height (cm)13.3
  • Motor hp0.13 to 0.29
  • DescriptionA-Mount Air-Driven Pump Drive, 500 to 8000 rpm, 5 to 11 scfm, 50 to 80 psi

Key Features

  • Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
  • Choose either an explosion-proof or air-powered model for safe pumping in hazardous areas
  • Hundreds of hours of dependable leakproof service

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat