Techne Fluidized sand bath systems; temperature range; 50-600°C; 240V; 2x 750W Heaters

Representative image only.
Techne Item # EW-01184-02
Fast heat-ups and accurate temperature control to 600°C
  • Dry, inert, nontoxic aluminum oxide ensures hazard-free operation
  • Safer alternative to high temperature oil and salt baths
  • Requires controller (1184-50) in order to be calibrated
  • Requires 20 lb (9 kg) of brown sand (01184-63)

PRODUCT OPTIONS

$4,800.00USD / Each

$285.00USD / Each

for an additional
$285.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Bath TypeSpecial Purpose Water Baths
  • Max Temperature (° C)600
  • Min Temperature (° C)50
  • Temperature Uniformity (° C)±1
  • Power (VAC)230
  • Power (Hz)50/60
  • DescriptionFluidized sand bath systems; temperature range; 50-600°C; 240V; 2x 750W Heaters

Includes

overspill flange, and power cord with plug.

Key Features

  • Dry, inert, nontoxic aluminum oxide ensures hazard-free operation
  • Safer alternative to high temperature oil and salt baths
  • Requires controller (1184-50) in order to be calibrated
  • Requires 20 lb (9 kg) of brown sand (01184-63)

More About this Item

Use these fluidized sand baths for your heat treating processes, temperature sensor calibrations, and electrical component testing. Baths use aluminum oxide fluidized by low-pressure air as a dry bath medium with excellent heat transfer properties. Inert, nonflammable aluminum oxide is safer to use than oil and will not corrode your sample vessels or produce toxic fumes. Vessels of all shapes can be easily immersed and safely removed from the bath, without the hazard and mess of hot oils. Make temperature adjustments via a control knob and reference dial. Baths require a dry air flow of approximately 3 cu ft/minute for model 01184-00 and -02 and 5 cu ft/minute for 01184-04 at about 3 psi pressure; order an air pump separately. All baths have a thermowell and an overspill flange. Note: not recommended for shape setting in laboratory research device manufacturing or for heating of reactors.

Controller (1184-50) sold seperately.

ACCESSORIES

REVIEWS

REVIEW SNAPSHOT®

by PowerReviews
Techne Fluidized sand bath systems; temperature range; 50-600?C; 240V; 2x 750W Heaters
 
3.0

(based on 1 review)

Reviewed by 1 customer

Displaying review 1

Back to top

(1 of 1 customers found this review helpful)

 
3.0

System with not enough sand

By 

from Santa Rosa, Ca

Pros

  • Accurate
  • Durable
  • Easy To Use
  • Works Properly

Cons

  • Not enough sand

Best Uses

  • In The Lab
  • Research

Comments about Techne Fluidized sand bath systems; temperature range; 50-600?C; 240V; 2x 750W Heaters:

Nice easy to use system, but there is not enough sand to fill the bath to the level of our liking. If you're going to pay several thousand dollars for the system, it should include enough sand to overfill the bath. Called tech support, but there is no recommendation to buy the sand.

Displaying review 1

Back to top

Q&A

Do you have questions about this product?

Get answers from real customers and in-house experts with AnswerBox.

3 Questions | 4 Answers
Displaying questions 1-3
  • Q:

    What are the heating up time from 20 C to 200 C for SN-01189-50 and SN-01184-02 sand baths?
    Asked on 3/26/2013 by sersan from Istanbul, Turkey

    2 answers

    • A:

      The heatup time to 200°C is approximately 30 minutes.

      Answered on 3/21/2017 by PJ from MN
    • STAFF EXPERT

      A:

      We have only specified the heat-up time from ambient to 350C (50 mins) and from ambient to 600C (70 mins) for the 01189-50. The heat-up time from ambient to 200C for the 01184-02 is 6 mins.

      Answered on 3/27/2013 by James from Technical Support
  • Q:

    Is the top of the cylinder open? I need to heat a quartz tube while it is connected to a manifold, and therefore the heating system must be open on the top so the tube can be partially submerged in the sand.
    Asked on 2/23/2012 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      The cylinder is open at the top, and the bath comes with the overspill flange accessory. That way you can fill the sand to the top without making a mess. You can purchase additional sand under part numbers 00152IM or 00257US.

      Answered on 2/28/2012 by Christina from Technical Support
  • Q:

    is the temperature controller required ?
    does the bath dial just read one to ten so that one must calibrate the dial with a thermocouple of some kind ?
    Asked on 10/7/2010 by Anonymous from drexel

    1 answer

    • Staff Reviewer

      A:

      Yes, for a complete system the bath, controller, and air pump must be purchased. The temperature controller uses a Type K thermocouple as to adjust and control temperature precisely. Yes, the sand bath just has an analog dial to adjust the temperature.

      Answered on 10/7/2010 by Jen from Technical Support
Displaying questions 1-3

Do you have a question about this product? 

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat