This item is now discontinued. We recommend the following as suggested replacements: EW-16101-89
Techne Stainless Steel Tank; 21 Liters capacity
Representative image only.
Techne Item # EW-01262-25
Five capacities available — ideal for a wide variety of vessel shapes and sizes Optional bridge allows you to easily convert tanks into heated water baths
Specifications & Description
  • Bath TypeTank Bath
  • Reservoir Capacity (L)26
  • Max Temperature (° C)250
  • Min Temperature (° C)0
  • Working Area Depth (in)7 1/16
  • Bath Opening Width (in)19 13/16
  • Bath Opening Length (in)11 3/16
  • Wetted Materials304 SS
  • DescriptionStainless Steel Tank; 21 Liters capacity
More About this Item
These versatile tanks are available in five capacities — up to 48.5 liters. Outer cases are constructed of baked enamel steel. Interiors are seamless 304 stainless steel (except for the 48.5 liter model which is welded). Carrying handles make tanks easier and safer to move. To convert these tanks into heated baths, simply order a high-stability immersion circulator from page 64 and a bridge from the table below. The optional 304 stainless steel flat cover prevents evaporation and reduces heat loss — helping to maintain a constant bath temperature. For extra headroom when bath is covered, gabled covers are available.
Use only with the high-stability immersion circulators.