This item is now discontinued. Please browse similar items
PVC Y-pattern globe valve with EPDM seat/seals; 1/2", socket
Representative image only.
Item # EW-01358-00
  • Excellent for throttling applications—globe valve design provides efficient flow control
  • Y-pattern reduces flow restriction over standard globe-type valves
  • Positive shut-off with minimal closing pressure
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/2" socket
  • Connection TypeSocket
  • Inlet Port1/2" socket
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC
  • DescriptionPVC Y-pattern globe valve with EPDM seat/seals; 1/2", socket
Key Features
  • Excellent for throttling applications—globe valve design provides efficient flow control
  • Y-pattern reduces flow restriction over standard globe-type valves
  • Positive shut-off with minimal closing pressure
  • Special semi-rigid seating disc eliminates vibration
More About this Item
Valves are designed for frequent operation with replaceable EPDM O-ring, seat and seals. All valves have a high-impact polypropylene hand wheel. Threaded connections are stainless steel reinforced. Note: not for use with compressed air or gas.