This item is now discontinued. Please browse similar items
PVC Y-pattern globe valve with EPDM seat/seals; 3" NPT(F)
Representative image only.
Item # EW-01358-28
  • Excellent for throttling applications—globe valve design provides efficient flow control
  • Y-pattern reduces flow restriction over standard globe-type valves
  • Positive shut-off with minimal closing pressure
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))3
  • Outlet Port3" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port3" NPT(F)
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC
  • DescriptionPVC Y-pattern globe valve with EPDM seat/seals; 3" NPT(F)
Key Features
  • Excellent for throttling applications—globe valve design provides efficient flow control
  • Y-pattern reduces flow restriction over standard globe-type valves
  • Positive shut-off with minimal closing pressure
  • Special semi-rigid seating disc eliminates vibration
More About this Item
Valves are designed for frequent operation with replaceable EPDM O-ring, seat and seals. All valves have a high-impact polypropylene hand wheel. Threaded connections are stainless steel reinforced. Note: not for use with compressed air or gas.