This item is now discontinued. Please browse similar items
PVC gate valve; 1" NPT(F), Buna N seals
Representative image only.
Item # EW-01359-04
  • Excellent for a variety of fluid-handling applications
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))1
  • Outlet Port1" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1" NPT(F)
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC
  • DescriptionPVC gate valve; 1" NPT(F), Buna N seals
Key Features
  • Excellent for a variety of fluid-handling applications
More About this Item
All valves are assembled with silicone-free, water-soluble lubricant.
Note: not for use with compressed air or gas.