This item is now discontinued. Please browse similar items
Advantec 6-Position Stirrer for Bath/Magnetic Stirrer System, 30mL to 12L, 120VAC
Representative image only.
Advantec Mfr # SRS266PA – Item # EW-02918-18
Efficient and Stable
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)115
  • Description6-Position Stirrer for Bath/Magnetic Stirrer System, 30mL to 12L, 120VAC
More About this Item
Magnetic stirrers can be used with our water bath or oil bath. This combination affords stable temperature control in various applications. The standard magnetic stirrer has smooth and clean design. The main body is made of GF-PBT, glass fiber-reinforced polybutylene terephthalate, which is heat and chemical resistant and durable.
The multiple stirring with powerful and precise control. Each stirring position has a powerful rare-earth magnet and electronic control circuit. It can also be controlled in a wide range of rotation, from low to high speed, simply by turning the switch at each stirring position.