Thermo Scientific Thermolyne HP11515B Explosion-Proof Hot Plate; 120VAC

Representative image only.
Thermo Scientific Thermolyne Mfr # HP11515B – Item # EW-03417-00
UL-Listed for Class 1, Group D hazardous locations
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Top Plate Length (in)6 1/8
  • Top Plate Width (in)6 1/8
  • Top Plate Length (cm)15.56
  • Top Plate Width (cm)15.56
  • Top Plate MaterialAluminum
  • Min Temperature (° F)100
  • Max Temperature (° F)428
  • Min Temperature (° C)38
  • Max Temperature (° C)220
  • Temperature Accuracy (° F)±9.9°
  • Temperature Accuracy (° C)±5.5°
  • Max Load Capacity (lbs)25
  • Max Load Capacity (kg)11.34
  • Length (in)7 5/8
  • Width (in)7
  • Height (in)3 1/4
  • Length (cm)19.3675
  • Width (cm)17.78
  • Height (cm)8.255
  • Power (VAC)120
  • Power (Hz)50/60
  • Power (watts)600
  • Power (amps)1.5
  • DescriptionExplosion-Proof Hot Plate; 120VAC
  • Warranty1 year
1 Year WarrantyUL Listed.

Key Features

  • 1 year warranty

More About this Item

Safely use this explosion-proof hot plate where flammable gases or vapors can produce explosive or ignitable mixtures—ideal for evaporating volatile organic solvents and heating lacquers.
Thermostat maintains temperature to within ±5.5degC. Added safety thermostat prevents temperatures from exceeding 243degC. Hot plate features a durable, corrosion-resistant steel case; cast-aluminum top plate accommodates metal vessels. Hot plate includes a ½'' conduit attachment and an 18'' lead wire for UL-required hookup (no plug). Optional dial thermometer fits directly into top plate to monitor top plate temperature—order separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat