This item is now discontinued. Please browse similar items

High-speed dispersion mixer, 0 to 20000 rpm, 230VAC, 50Hz

Representative image only.
Item # EW-04201-07
Efficiently disperse and mix hard-to-wet fine powders and settling solids
  • Obtain reproducible results, which can be compared exactly
  • Simple and quiet operation


Specifications & Description

  • Min Speed (rpm)0
  • Max Speed (rpm)20000
  • Power (Phase)1
  • Power (VAC)230
  • Power (Hz)50
  • Motor hp3/4
  • DescriptionHigh-speed dispersion mixer, 0 to 20000 rpm, 230VAC, 50Hz

Key Features

  • Obtain reproducible results, which can be compared exactly
  • Simple and quiet operation

More About this Item

Dispersion devices are used to incorporate extremely fine solid particles into the surrounding medium, such that the small particles do not settle under the force of gravity. Efficient dispersion is easy to obtain. First, the liquid is put into the container. Then, under moderate speed, the small particles are added. The impeller speed can be increased until the doughnut flow pattern is detected at a higher rpm. After this premixing steps the dispersion is carried out at high peripheral speeds that correlate to the tip speeds used in production by maintaining the doughnut. When dispersing small quantities, use high rotational speeds to match production results.
High-Speed Dispersion Mixers have cruise control to maintain a constant speed even when viscosity changes for repeatable results. Model 04201-00 is a low cost solution for smaller quantities. Models 04201-02 and –03 are ideal for dispersion tasks in the lab and have heavy-duty clamps to tightly secure samples during maximum speeds.
High-Speed Mixers include motor, control box, support stand, and four impellers in the following sizes: 20, 25, 30, and 40 mm.
Reversible Dispersion Mixers allow you to change mixing directions. Downward “vortex” flow and upward “umbrella” flow ensure thoroughly mixed ingredients from center to wall and from top to bottom of the tank. Model 50401-01 has a solid-state, NEMA 4 control box that connects to the motor via a 5-ft cord with waterproof plug. Controller features on/off switch; pilot light; reset button; speed control dial; and mode selector (downward, stop, and upward). Choose air-drive model 50401-10 for mixing in hazardous atmospheres. The 1/2 hp air-drive motor is ideal for mixing solvents and volatile liquids. The motor accepts air line pressures up to 150 psig at 25 cfm.
Reversible Mixers include support stand with anti-tip base and post providing 183/4" vertical travel, 6" horizontal travel, 303/304 SS shaft, pitched rotor, adjustable baffle plate, pressure regulator, oiler and reversing valve and 360° rotation.




Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat