Thermo Scientific Nalgene 3665-0500 graduated polymethylpentene cylinder, 500 mL

Representative image only.
Thermo Scientific Nalgene Mfr # 3665-0500 – Item # EW-06134-65
Easy-to-read raised graduations
  • Chemical and heat resistant
  • Large pour spout
  • Autoclavable
  • 1 year warranty

PRODUCT OPTIONS

$39.50USD / Pkg of 1

BUY MORE & SAVE

  • 1-5$39.50USD / Pkg of 1
  • 6-29$35.55USD / Pkg of 1
  • 30-119$33.58USD / Pkg of 1
  • 120+$31.60USD / Pkg of 1

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPolymethylpentene
  • Capacity (mL)500
  • Graduation Interval (mL)5
  • Tolerance (mL)±2.6
  • Descriptiongraduated polymethylpentene cylinder, 500 mL
  • Warranty1 year
1 Year Warranty

Key Features

  • Chemical and heat resistant
  • Large pour spout
  • Autoclavable
  • 1 year warranty

More About this Item

Transparent polymethylpentene (PMP) cylinders with large pour spout offer good chemical and heat resistance. Easy-to-read, raised graduations allow precise measuring.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat